Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-methoxytoluene 98.0+%, TCI America™
SDP

Catalog No. B35261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35261G 1 g
B35265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35261G Supplier TCI America Supplier No. B35261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29578-83-4
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD08061916
InChI Key AOEVRCZZWJWKPG-UHFFFAOYSA-N
Synonym 3-Bromo-5-methylanisole
PubChem CID 10679554
IUPAC Name 1-bromo-3-methoxy-5-methylbenzene
SMILES CC1=CC(=CC(=C1)Br)OC

Specifications

Boiling Point 122°C
Quantity 1 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-methoxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.