Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-nitrobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. B53335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53335G 5 g
B533325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53335G Supplier TCI America Supplier No. B53335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 630125-49-4
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.01
MDL Number MFCD03788719
InChI Key YHTVYRKVFAFVLP-UHFFFAOYSA-N
Synonym 3-bromo-5-nitrobenzotrifluoride, 1-bromo-3-nitro-5-trifluoromethyl benzene, 3-bromo-5-trifluoromethylnitrobenzene, 3-nitro-5-bromobenzotrifluoride, benzene, 1-bromo-3-nitro-5-trifluoromethyl, 5-bromo-3-trifluoromethyl nitrobenzene, 1-bromo-3-nitro-5-trifluoromethyl-benzene, 3-bromo-5-nitrotrifluorotoluene, 3-bromo-1-nitro-5-trifluoromethyl benzene, zlchem 416
PubChem CID 7015411
IUPAC Name 1-bromo-3-nitro-5-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(Br)=CC(=C1)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 270.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.