Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-nitropyridine, 97%, Thermo Scientific™

Catalog No. AAH33531MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33531MD 250 mg
AAH3353103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33531MD Supplier Thermo Scientific Chemicals Supplier No. H33531MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15862-30-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 202.995
MDL Number MFCD04114216
InChI Key WHYJVHQKKJHXTB-UHFFFAOYSA-N
Synonym 3-bromo-5-nitro-pyridine, pyridine, 3-bromo-5-nitro, acmc-1bqk8, 3-bromo-5-nitropyridine?, pyridine,3-bromo-5-nitro, 3-bromanyl-5-nitro-pyridine
PubChem CID 2762898
IUPAC Name 3-bromo-5-nitropyridine
SMILES C1=C(C=NC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 111°C to 115°C
Quantity 250 mg
Formula Weight 203
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-nitropyridine
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 677-831-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.