Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-phenylpyridine 98.0+%, TCI America™
SDP

Catalog No. B5151200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5151200MG 200 mg
B51511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5151200MG Supplier TCI America Supplier No. B5151200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142137-17-5 | C11H8BrN | 234.096 g/mol

Chemical Identifiers

CAS 142137-17-5
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD04114223
InChI Key AWCQJXPOCRXHNK-UHFFFAOYSA-N
PubChem CID 2762904
IUPAC Name 3-bromo-5-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CC(=CN=C2)Br

Specifications

Melting Point 48°C
Color White
Boiling Point 101°C
Quantity 200 mg
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.