Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B49745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49745G 5 g
B49741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49745G Supplier TCI America Supplier No. B49745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 477535-41-4
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.02
MDL Number MFCD08059506
InChI Key PCRLZGCXLNNMFL-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzaldehyde, benzaldehyde, 3-bromo-5-trifluoromethyl, 3-trifluoromethyl-5-bromobenzaldehyde, 3-bromo-5-trifluoromethyl-benzaldehyde, pubchem16131, acmc-209kad, ksc496e1r, 3-bromo-5-formylbenzotrifluoride, 5-bromo-3-trifluoromethyl benzaldehyde
PubChem CID 16115438
IUPAC Name 3-bromo-5-(trifluoromethyl)benzaldehyde
SMILES FC(F)(F)C1=CC(Br)=CC(C=O)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 253.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.