Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-6,9-diphenylcarbazole 98.0+%, TCI America™
SDP

Catalog No. B47955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47955G 5 g
B47951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47955G Supplier TCI America Supplier No. B47955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1160294-85-8
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.30
MDL Number MFCD20257783
InChI Key JBEQRROTSDUCGC-UHFFFAOYSA-N
PubChem CID 57746295
IUPAC Name 3-bromo-6,9-diphenyl-9H-carbazole
SMILES BrC1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 5 g
Formula Weight 398.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-6,9-diphenylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.