Learn More
Medchemexpress LLC 3-bromo-7-nitroindazole | 74209-34-0 | MFCD00159910 | 98.1% | 242.03 g/mol | C7H4BrN3O2 | 100 MG
SDP
Catalog No. 5000212480
Encompass_Preferred
3-Bromo-7-nitroindazole is a research-grade small molecule that potently and selectively inhibits neuronal nitric oxide synthase (nNOS). Supplied as a solid powder, it is used in preclinical in vitro and in vivo studies to probe nitric oxide-mediated signaling in neurological and metabolic disease models.
  • Potent, selective nNOS inhibitor.
  • High purity (~98.1%).
  • Solid powder form for in vitro and in vivo use.
  • Available in small research pack sizes, including 100 MG.
  • Recommended storage for stability: powder -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-12480 Supplier Medchemexpress LLC Supplier No. HY101175100MG
3-Bromo-7-nitroindazole is a research-grade small molecule that potently and selectively inhibits neuronal nitric oxide synthase (nNOS). Supplied as a solid powder, it is used in preclinical in vitro and in vivo studies to probe nitric oxide-mediated signaling in neurological and metabolic disease models.
  • Potent, selective nNOS inhibitor.
  • High purity (~98.1%).
  • Solid powder form for in vitro and in vivo use.
  • Available in small research pack sizes, including 100 MG.
  • Recommended storage for stability: powder -20°C (up to 3 years) or 4°C (up to 2 years).