Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-9-(4-bromophenyl)-9H-carbazole 97.0+%, TCI America™
SDP

Catalog No. B51075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51075G 5 g
B51071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51075G Supplier TCI America Supplier No. B51075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1226860-66-7
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.10
MDL Number MFCD30063199
InChI Key ZUNLPNWEQKBULG-UHFFFAOYSA-N
PubChem CID 66797808
IUPAC Name 3-bromo-9-(4-bromophenyl)-9H-carbazole
SMILES BrC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 401.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-(4-bromophenyl)-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.