Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-9-hexyl-9H-carbazole 99.0+%, TCI America™
SDP

Catalog No. B56085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56085G 5 g
B56081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56085G Supplier TCI America Supplier No. B56085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156972-74-6
Molecular Formula C18H20BrN
Molecular Weight (g/mol) 330.27
MDL Number MFCD30562084
InChI Key HYUCKXZMLJFXSS-UHFFFAOYSA-N
PubChem CID 71354044
IUPAC Name 3-bromo-9-hexyl-9H-carbazole
SMILES CCCCCCN1C2=C(C=CC=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 48°C
Quantity 5 g
Formula Weight 330.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-hexyl-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.