Learn More
eMolecules 3-Bromo-biphenyl-4-ylamine | 41738-70-9 | MFCD00034057 | 5g
Supplier: eMolecules 309209743
J & W PharmLab LLC | 3-Bromo-biphenyl-4-ylamine | 5g | 309209743 | 20R1582 | 96.000 | 41738-70-9 | MFCD00034057 | 248.123 | C12H10BrN
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool