Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Bromo-DL-phenylalanine | 30163-20-3 | 244.09 | 100 G
SDP

Catalog No. 5000449044
Encompass_Preferred

3-Bromo-DL-phenylalanine is a derivative of phenylalanine, an amino acid. Amino acids and their derivatives are widely utilized commercially as ergogenic supplements due to their ability to influence anabolic hormone secretion, provide energy during exercise, enhance mental performance under stress, and prevent exercise-induced muscle damage. These compounds are recognized as beneficial ergogenic dietary substances. It has a molecular formula of C9H10BrNO2, appears as a white to off-white solid, and has a molecular weight of 244.09.

  • Molecular formula: C9H10BrNO2
  • Appearance: White to off-white solid
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 2 mg/mL (8.19 mM; with ultrasonic and warming to 60°C)

Catalog No. 50-004-49044 Supplier Medchemexpress LLC Supplier No. HYW003225100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Bromo-DL-phenylalanine is a derivative of phenylalanine, an amino acid. Amino acids and their derivatives are widely utilized commercially as ergogenic supplements due to their ability to influence anabolic hormone secretion, provide energy during exercise, enhance mental performance under stress, and prevent exercise-induced muscle damage. These compounds are recognized as beneficial ergogenic dietary substances. It has a molecular formula of C9H10BrNO2, appears as a white to off-white solid, and has a molecular weight of 244.09.

  • Molecular formula: C9H10BrNO2
  • Appearance: White to off-white solid
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 2 mg/mL (8.19 mM; with ultrasonic and warming to 60°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.