Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH60571MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60571MD 250 mg
AAH6057103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60571MD Supplier Thermo Scientific Chemicals Supplier No. H60571MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1024483-64-4
Molecular Formula C14H11BrF3NO2S
Molecular Weight (g/mol) 394.206
MDL Number MFCD22683137
InChI Key RZKLDSNLHNWKFJ-UHFFFAOYSA-N
Synonym 3-bromo-n-4-trifluoromethyl phenyl methyl benzenesulfonamide, 3-bromo-n-4-trifluoromethyl benzyl benzenesulfonamide
PubChem CID 4986333
IUPAC Name 3-bromo-N-[[4-(trifluoromethyl)phenyl]methyl]benzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CC=C(C=C2)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 394.21
Percent Purity 97%
Chemical Name or Material 3-Bromo-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.