Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromobenzenesulfonyl fluoride

Catalog No. ALD002121G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD002121G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD002121G Supplier Sigma Aldrich Supplier No. ALD002121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4-hexadiene, mixture of isomers, is a conjugated diene
  • The interaction between palladium chloride and 2,4-hexadiene has been reported to proceed in a stereospecific manner
  • The photodissociation spectra of 2,4-hexadiene isomers have been recorded
  • The polymers obtained by the polymerization of 2,4-hexadiene isomers are reported to have the trans-1,4 structure

Specifications

CAS 454-65-9
Density 1.7355 (at 25°C)
Molecular Formula C6H4BrFO2S
Refractive Index n/D 1.5392
Linear Formula C6H4BrFO2S
MDL Number MFCD22053310
Quantity 10 g
Synonym 3-Bromobenzenesulfonyl fluoride
Molecular Weight (g/mol) 239.06
Percent Purity 95% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.