Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzenethiol 98.0+%, TCI America™
SDP

Catalog No. B23485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23485G 5 g
B234825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23485G Supplier TCI America Supplier No. B23485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6320-01-0
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00009603
InChI Key HNGQQUDFJDROPY-UHFFFAOYSA-N
Synonym 3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, #
PubChem CID 80597
IUPAC Name 3-bromobenzene-1-thiol
SMILES SC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 g
UN Number 1760
Formula Weight 189.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzenethiol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.