Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobutyronitrile 98.0+%, TCI America™
SDP

Catalog No. B14525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14525G 5 g
B145225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14525G Supplier TCI America Supplier No. B14525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20965-20-2
Molecular Formula C4H6BrN
Molecular Weight (g/mol) 148.003
MDL Number MFCD00039488
InChI Key MBXSHBIQMDKTEW-UHFFFAOYSA-N
Synonym 2-Bromopropyl Cyanide
PubChem CID 549273
IUPAC Name 3-bromobutanenitrile
SMILES CC(CC#N)Br

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 3276
Formula Weight 148.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobutyronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.