Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt 98.0+%, TCI America™
SDP

Catalog No. B11255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11255G 5 g
B112525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11255G Supplier TCI America Supplier No. B11255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14575-84-9
Molecular Formula C10H18BrNO4S
Molecular Weight (g/mol) 328.221
MDL Number MFCD00167438
InChI Key GFBVBBRNPGPROZ-ZEMXJPTRSA-N
Synonym d-3-bromocamphor-8-sulfonic acid ammonium salt, +-3-bromocamphor-8-sulfonic acid ammonium salt, 1r-endo,anti-+-3-bromocamphor-8-sulfonic acid ammonium salt, 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonic acid amine, ammonium 1r,3s,4s,7r-3-bromo-1,7-dimethyl-2-oxobicyclo 2.2.1 heptan-7-yl methanesulfonate
PubChem CID 71306887
IUPAC Name azane;[(1S,2S,4R,7R)-2-bromo-4,7-dimethyl-3-oxo-7-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC12CCC(C1(C)CS(=O)(=O)O)C(C2=O)Br.N

Specifications

Quantity 5 g
Formula Weight 328.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-3-Bromocamphor-8-sulfonic Acid Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.