Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocumene 98.0+%, TCI America™
SDP

Catalog No. B28755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28755G 5 g
B287525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28755G Supplier TCI America Supplier No. B28755G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5433-01-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD01318112
InChI Key GBSGGFCCQZUXNB-UHFFFAOYSA-N
Synonym 1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene
PubChem CID 138499
IUPAC Name 1-bromo-3-propan-2-ylbenzene
SMILES CC(C)C1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 199.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromocumene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.