Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromofuran, 98%, stabilized with CaCO3

Catalog No. AC175260100
Change view
Click to view available options
Quantity:
2.5 mL
10 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175260100 10 mL Glass bottle
AC175260025 2.5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC175260100 Supplier Thermo Scientific Chemicals Supplier No. 175260100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22037-28-1 | C4H3BrO | 146.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.6350g/mL
Boiling Point 103.0°C (750.0 mmHg)
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Quantity 10 mL
Specific Gravity 1.635
Stabilizer copper
Formula Weight 146.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromofuran, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 244-744-6
Recommended Storage −20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.