Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™

Catalog No. CC25410EA
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC25410EA 10 g
CC25410DA 1 g
CC25410DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC25410EA Supplier Thermo Scientific Chemicals Supplier No. CC25410EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4926-47-0
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
InChI Key APYSHMNJHJRIDR-UHFFFAOYSA-N
PubChem CID 327951
IUPAC Name 3-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=C(N2C=C1)Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Synonym 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromoimidazo[1, 2-a]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.