Learn More
Medchemexpress LLC 3-bromoisobutyric acid | 56970-78-6 | MFCD01310530 | 95.0% | 167.00 g/mol | C4H7BrO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01533210MM/1ML
Encompass_Preferred
Captopril EP Impurity D (3-bromoisobutyric acid) is an analytical reference impurity of captopril provided as a 10 mM solution in DMSO for method development, impurity profiling, and pharmaceutical quality-control testing. CAS 56970-78-6; molecular formula C4H7BrO2; reported purity 95.0%.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for analytical method development and impurity profiling.
  • Useful as a reference standard in pharmaceutical quality control.
  • Supplied with a documented purity specification for traceability.
  • Enables accurate identification and quantification by LC/MS and related techniques.
Catalog No. 50-003-02413