Learn More
Medchemexpress LLC 3-bromoisobutyric acid | 56970-78-6 | MFCD01310530 | >97.0% | 167.00 g/mol | C4H7BrO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0153325G
Encompass_Preferred
Captopril EP Impurity D (3-bromoisobutyric acid) is an analytical reference standard used for identification and quantification of a captopril-related impurity in pharmaceutical analysis. Supplied as a colorless to light-yellow liquid, it supports method development, quality control, and impurity profiling in laboratory and regulatory environments.
  • Analytical reference standard for impurity identification and quantification.
  • Provided as a liquid suitable for analytical preparation.
  • High purity for reliable assay and method validation.
  • Available in multiple pack sizes to suit laboratory needs.
  • Accompanied by documentation such as datasheet, COA, and SDS for regulatory compliance.
Catalog No. 50-003-02414