Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Bromomethyl)-5-chlorobenzo[b]thiophene 98.0+%, TCI America™
SDP

Catalog No. B31471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31471G 1 g
B31475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31471G Supplier TCI America Supplier No. B31471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1198-51-2
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.56
MDL Number MFCD00173769
InChI Key FKQSFVITUNJLCY-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553
PubChem CID 2779857
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
SMILES ClC1=CC=C2SC=C(CBr)C2=C1

Specifications

Melting Point 131°C
Quantity 1 g
UN Number 3261
Formula Weight 261.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Bromomethyl)-5-chlorobenzo[b]thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.