Learn More
Accela Chembio Inc 3-(bromomethyl)pyridine Hydrobromide | 5g | 4916-55-6 | MFCD01321314 | 95+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50249756
Encompass_Preferred
3-(bromomethyl)pyridine Hydrobromide | 5g | 4916-55-6 | MFCD01321314 | 95+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-249-756 Supplier Accela Chembio Inc Supplier No. SY0023025G
3-(bromomethyl)pyridine Hydrobromide | 5g | 4916-55-6 | MFCD01321314 | 95+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4