Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromooxetane

Catalog No. 756962500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-696-2500MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-696-2500MG Supplier Sigma Aldrich Supplier No. 756962500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Pyridinemethanol, an aromatic primary alcohol, is the key moiety of many bio-active and industrially important compounds.3-Pyridinemethanol can undergo aerobic photo-oxidation in the presence of catalytic amount of hydrobromic acid to form nicotinic acid.

Specifications

CAS 39267-79-3
Density 1.651 g/mL (at 25°C)
Molecular Formula C3H5BrO
Refractive Index n20/D 1.484
Linear Formula C3H5BrO
MDL Number MFCD08544402
Quantity 100 g
Molecular Weight (g/mol) 136.98
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.