Learn More
Sigma Aldrich (3-Bromophenyl)triphenylsilane
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-(dimethylamino)pyridine is a clear, colorless to light yellow liquid.2-(dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt
- It is used as a model substrate in the preparation of chelate carbene via cyclometalation, h2 loss and reversible α-elimination
Specifications
Specifications
| Molecular Formula | C24H19BrSi |
| Linear Formula | C24H19BrSi |
| MDL Number | MFCD28098175 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 415.4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.