Learn More
Medchemexpress LLC 3-Bromophenylacetic acid | 1878-67-7 | MFCD00004330 | 215.05 | 25 G
SDP
Catalog No. 5000449981
Encompass_Preferred
3-Bromophenylacetic acid, also known as 2-(3-Bromophenyl)acetic acid, is a biochemical reagent. It serves as a valuable biological material and organic compound for various life science-related research applications. This chemical compound is essential for researchers in need of a high-purity reagent for their experiments.
  • Used as a biochemical reagent in life science research
  • Suitable as a biological material or organic compound
  • Ensure container is tightly sealed and stored in a cool, well-ventilated area
  • Protect from direct sunlight and sources of ignition
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-49981 Supplier Medchemexpress LLC Supplier No. 25GHYW004766
3-Bromophenylacetic acid, also known as 2-(3-Bromophenyl)acetic acid, is a biochemical reagent. It serves as a valuable biological material and organic compound for various life science-related research applications. This chemical compound is essential for researchers in need of a high-purity reagent for their experiments.
  • Used as a biochemical reagent in life science research
  • Suitable as a biological material or organic compound
  • Ensure container is tightly sealed and stored in a cool, well-ventilated area
  • Protect from direct sunlight and sources of ignition
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month