Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B28905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2890-5G 5 g
B2890-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2890-5G Supplier TCI America Supplier No. B28905G
Add to Cart
Add to Cart

Specifications

CAS 89598-96-9
Melting Point 168°C
Color White-Yellow
Molecular Formula C6H6BBrO2
MDL Number MFCD00239386
Quantity 5 g
Synonym 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758
InChI Key AFSSVCNPDKKSRR-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)Br)(O)O
IUPAC Name (3-bromophenyl)boronic acid
Molecular Weight (g/mol) 200.826
PubChem CID 2734318
Formula Weight 200.83
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.