Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Bromopropan-1-ol | 627-18-9 | MFCD00002942 | 138.99 | 25 G
SDP

Catalog No. 5000449932
Encompass_Preferred

3-Bromopropan-1-ol is an endogenous metabolite that has been observed to be significantly upregulated during the estrus period in buffaloes. This compound can be utilized in buffalo estrus research.

  • Endogenous metabolite
  • Applicable in buffalo estrus research
  • High purity of 99.07%
  • Stable in pure form for up to 3 years at -20°C or 2 years at 4°C
  • Solution stability up to 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-49932 Supplier Medchemexpress LLC Supplier No. 25GHYW007337
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Bromopropan-1-ol is an endogenous metabolite that has been observed to be significantly upregulated during the estrus period in buffaloes. This compound can be utilized in buffalo estrus research.

  • Endogenous metabolite
  • Applicable in buffalo estrus research
  • High purity of 99.07%
  • Stable in pure form for up to 3 years at -20°C or 2 years at 4°C
  • Solution stability up to 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.