Learn More
Medchemexpress LLC 3-Bromopropan-1-ol-d6 | 284474-43-7 | 145.03 | 10 MG
SDP
Catalog No. 5000460336
Encompass_Preferred
3-Bromopropan-1-ol-d6 is a deuterium-labeled form of 3-Bromopropan-1-ol, an endogenous metabolite. It is significantly upregulated during the estrus period in buffaloes and can be utilized in buffalo estrus research. Deuterium substitution, involving stable heavy isotopes, is incorporated into drug molecules primarily as tracers for quantitation during drug development, showing potential to influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-60336 Supplier Medchemexpress LLC Supplier No. HYW007337S10MG
3-Bromopropan-1-ol-d6 is a deuterium-labeled form of 3-Bromopropan-1-ol, an endogenous metabolite. It is significantly upregulated during the estrus period in buffaloes and can be utilized in buffalo estrus research. Deuterium substitution, involving stable heavy isotopes, is incorporated into drug molecules primarily as tracers for quantitation during drug development, showing potential to influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.