Learn More
Medchemexpress LLC 3-Bromopropan-1-ol-d6 | 284474-43-7 | 145.03 | 50 MG
SDP
Catalog No. 5000460379
Encompass_Preferred
This product is a deuterium-labeled version of 3-Bromopropan-1-ol, an endogenous metabolite. It has been observed to be significantly upregulated in buffaloes during the estrus period. This makes it a valuable tool for research related to buffalo estrus.
  • Tracer applications
  • Quantitative analysis
  • Isotope labeling
  • High purity
  • Research use only
Catalog No. 50-004-60379 Supplier Medchemexpress LLC Supplier No. HYW007337S50MG
This product is a deuterium-labeled version of 3-Bromopropan-1-ol, an endogenous metabolite. It has been observed to be significantly upregulated in buffaloes during the estrus period. This makes it a valuable tool for research related to buffalo estrus.
  • Tracer applications
  • Quantitative analysis
  • Isotope labeling
  • High purity
  • Research use only