Learn More
Medchemexpress LLC 3-Bromopropan-1-ol-d6 | 284474-43-7 | MFCD01075414 | 145.03 | 100 MG
SDP
Catalog No. 5000460357
Encompass_Preferred
3-Bromopropan-1-ol-d6 is the deuterium labeled form of 3-Bromopropan-1-ol, an endogenous metabolite significantly upregulated during the estrus period in buffaloes. This compound can be utilized in buffalo estrus research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-60357 Supplier Medchemexpress LLC Supplier No. HYW007337S100MG
3-Bromopropan-1-ol-d6 is the deuterium labeled form of 3-Bromopropan-1-ol, an endogenous metabolite significantly upregulated during the estrus period in buffaloes. This compound can be utilized in buffalo estrus research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.