Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-bromopropane-1,2-diol-d5 | 1246820-48-3 | 160.02 g·mol⁻¹ | C3H2D5BrO2 | 1 MG
SDP

Catalog No. 5000299774
Encompass_Preferred

3-Bromopropane-1,2-diol-d5 is the deuterium-labeled analogue of 3-bromopropane-1,2-diol, provided for use as an internal standard or tracer in analytical chemistry workflows such as NMR, GC-MS, and LC-MS. It is supplied in small milligram quantities for research and quantitative applications.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Useful as a tracer for metabolic and stability studies.
  • Available in milligram pack sizes suitable for analytical workflows.
  • Molecular formula C3H2D5BrO2; molecular weight 160.02 g·mol⁻¹.
  • CAS number 1246820-48-3.

Catalog No. 50-002-99774 Supplier Medchemexpress LLC Supplier No. HYW017310S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Bromopropane-1,2-diol-d5 is the deuterium-labeled analogue of 3-bromopropane-1,2-diol, provided for use as an internal standard or tracer in analytical chemistry workflows such as NMR, GC-MS, and LC-MS. It is supplied in small milligram quantities for research and quantitative applications.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS methods.
  • Useful as a tracer for metabolic and stability studies.
  • Available in milligram pack sizes suitable for analytical workflows.
  • Molecular formula C3H2D5BrO2; molecular weight 160.02 g·mol⁻¹.
  • CAS number 1246820-48-3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.