Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopropionic acid, 97%

Catalog No. AC325770100
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325770100 10 g Glass Bottle
AC325771000 100 g Plastic bottle
AC325775000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC325770100 Supplier Thermo Scientific Chemicals Supplier No. 325770100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 590-92-1 | C3H5BrO2 | 152.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-92-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00002763
InChI Key DHXNZYCXMFBMHE-UHFFFAOYSA-N
Synonym 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
PubChem CID 11553
IUPAC Name 3-bromopropanoic acid
SMILES OC(=O)CCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 63°C
Color White to Yellow
Boiling Point 140°C to 142°C (45.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Linear Formula BrCH2CH2CO2H
Quantity 10 g
Beilstein 02, 256
Merck Index 15, 1444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chcl3 and benzene
Formula Weight 152.98
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Bromopropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 209-694-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.