Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromopropionitrile

Catalog No. 109231100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-923-1100G 100 g
10-923-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-923-1100G Supplier Sigma Aldrich Supplier No. 109231100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile
  • 3-bromopropionitrile was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-celite

Specifications

CAS 2417-90-5
Density 1.615 g/mL (at 25°C (literature))
Boiling Point 76°C to 78°C (10 mmHg)
Molecular Formula C3H4BrN
Refractive Index n20/D 1.481 (literature)
Linear Formula BrCH2CH2CN
MDL Number MFCD00001951
Quantity 100 g
Molecular Weight (g/mol) 133.97
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.