Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopropionitrile 97.0+%, TCI America™
SDP

Catalog No. B176225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B176225G Supplier TCI America Supplier No. B176225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2417-90-5
Molecular Formula C3H4BrN
Molecular Weight (g/mol) 133.976
MDL Number MFCD00001951
InChI Key CQZIEDXCLQOOEH-UHFFFAOYSA-N
Synonym 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile
PubChem CID 17020
IUPAC Name 3-bromopropanenitrile
SMILES C(CBr)C#N

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
UN Number 3276
Formula Weight 133.98
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromopropionitrile
RTECSNumber UG1050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.