Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Bromopropyl)Trichlorosilane

Catalog No. 4378085ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-780-85ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 43-780-85ML Supplier Sigma Aldrich Supplier No. 4378085ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Bromopropyl)trichlorosilane can be used: To prepare coupling agent 3-iodopropyltrichlorosilane (3-IC3H6SiCl3), which is employed for the preparation of self-assembled nonlinear optically (NLO) active chromophoric multilayers. As a reagent for the surface modification of silica nanofiber films and silica particles.

Specifications

CAS 10060-10-3
Molecular Formula CeF4
Linear Formula CeF4
Refractive Index n20/D 1.494
Quantity 5 mL
MDL Number MFCD00049422
Molecular Weight (g/mol) 216.11 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.