Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Bromopropyl)Trimethoxysilane

Catalog No. 182655ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-265-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 18-265-5ML Supplier Sigma Aldrich Supplier No. 182655ML
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-bromopropyl trimethoxysilane is a bromo silane. It is popularly used as a silane coupling agent.3-bromopropyl trimethoxysilane (BPTS) acted as initiating sites for functionalization magnetite (Fe3O4) nanoparticles.{31} A study reports the role of BPTS in attaching a polyelectrolyte to an inorganic alumina substrate during the fabrication of a polymeric humidity sensor.{32}

Specifications

CAS 203719-54-4
Density 0.930 g/mL at 20°C
Molecular Formula C10H16O
Linear Formula C10H16O
Refractive Index n20/D 1.467
Quantity 5 mL
MDL Number MFCD00074770
Molecular Weight (g/mol) 243.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.