Learn More
Medchemexpress LLC 3-Bromopyridin-2-ol | 13466-43-8 | 174.00 | 1 ML
SDP
Catalog No. 5000446692
Encompass_Preferred
3-Bromopyridin-2-ol is a biochemical reagent suitable for life science research. It functions as a biological material or organic compound, intended strictly for research purposes.
  • High purity (99.62%)
  • White to light brown solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Can be formulated for in vivo studies using various co-solvents
  • Stable for extended periods under recommended storage conditions
  • Used in biochemical assay applications
Catalog No. 50-004-46692 Supplier Medchemexpress LLC Supplier No. HYW00304510MM/1ML
3-Bromopyridin-2-ol is a biochemical reagent suitable for life science research. It functions as a biological material or organic compound, intended strictly for research purposes.
  • High purity (99.62%)
  • White to light brown solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Can be formulated for in vivo studies using various co-solvents
  • Stable for extended periods under recommended storage conditions
  • Used in biochemical assay applications