Learn More
Medchemexpress LLC 3-Bromopyridin-2-ol | 13466-43-8 | 174.00 | 25 G
SDP
Catalog No. 5000449420
Encompass_Preferred
3-Bromopyridin-2-ol is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. This product is intended for research use only and is not sold to patients. It is also identified as a laboratory chemical for the manufacture of substances.
  • High purity of 99.6%
  • Appears as an off-white to light brown solid
  • Soluble in DMSO (100 mg/mL)
  • Stable under recommended storage conditions
Catalog No. 50-004-49420 Supplier Medchemexpress LLC Supplier No. 25GHYW003045
3-Bromopyridin-2-ol is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. This product is intended for research use only and is not sold to patients. It is also identified as a laboratory chemical for the manufacture of substances.
  • High purity of 99.6%
  • Appears as an off-white to light brown solid
  • Soluble in DMSO (100 mg/mL)
  • Stable under recommended storage conditions