Learn More
AdipoGen 3-Bromopyruvic acid
SDP
Catalog No. 502562231
Encompass_Preferred
Chemical. CAS 1113-59-3. Formula C3H3BrO3. MW 167. Synthetic. Cell-permeable pyruvic acid derivative. Hexokinase II HK2 inhibitor. Useful agent for immunometabolism research. Antitumor agent. Inhibits the rate limiting step of glycolysis and effectively suppress tumor survival in vitro and in vivo. Displays potent cytotoxic activity against cancer cells with mitochondrial respiratory defects and cells in hypoxic environment. Antifungal agent anticryptococcal. Also used as affinity label for cysteine residues and cross-linker between nucleic acids and proteins.
Catalog No. 50-256-2231 Supplier AdipoGen Supplier No. AGCR13682G001
Chemical. CAS 1113-59-3. Formula C3H3BrO3. MW 167. Synthetic. Cell-permeable pyruvic acid derivative. Hexokinase II HK2 inhibitor. Useful agent for immunometabolism research. Antitumor agent. Inhibits the rate limiting step of glycolysis and effectively suppress tumor survival in vitro and in vivo. Displays potent cytotoxic activity against cancer cells with mitochondrial respiratory defects and cells in hypoxic environment. Antifungal agent anticryptococcal. Also used as affinity label for cysteine residues and cross-linker between nucleic acids and proteins.