Learn More
Medchemexpress LLC 3-Bromopyruvic acid | 1113-59-3 | 98.0% | 166.96 | 1 ML
SDP
Catalog No. 5000402038
Encompass_Preferred
3-Bromopyruvic acid is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis through interfering with glycolysis. This compound also induces autophagy by stimulating ROS formation in breast cancer cells, and exhibits antimicrobial activities.
  • Potent hexokinase (HK)-II inhibitor with high tumor selectivity.
  • Inhibits cell growth and induces apoptosis through interfering with glycolysis.
  • Induces autophagy by stimulating ROS formation in breast cancer cells.
  • Exhibits antimicrobial activities.
Catalog No. 50-004-02038 Supplier Medchemexpress LLC Supplier No. HY1999210MM/1ML
3-Bromopyruvic acid is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis through interfering with glycolysis. This compound also induces autophagy by stimulating ROS formation in breast cancer cells, and exhibits antimicrobial activities.
  • Potent hexokinase (HK)-II inhibitor with high tumor selectivity.
  • Inhibits cell growth and induces apoptosis through interfering with glycolysis.
  • Induces autophagy by stimulating ROS formation in breast cancer cells.
  • Exhibits antimicrobial activities.