Learn More
Medchemexpress LLC 3-Bromopyruvic acid | 1113-59-3 | 98.0% | 166.96 | 25 G
SDP
Catalog No. 5000402838
Encompass_Preferred
3-Bromopyruvate is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis by interfering with glycolysis, and induces autophagy by stimulating ROS formation in breast cancer cells. It also exhibits antimicrobial activities.
  • Analogue of pyruvate
  • Potent hexokinase (HK)-II inhibitor with high tumor selectivity
  • Inhibits cell growth
  • Induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities
Catalog No. 50-004-02838 Supplier Medchemexpress LLC Supplier No. 25GHY19992
3-Bromopyruvate is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis by interfering with glycolysis, and induces autophagy by stimulating ROS formation in breast cancer cells. It also exhibits antimicrobial activities.
  • Analogue of pyruvate
  • Potent hexokinase (HK)-II inhibitor with high tumor selectivity
  • Inhibits cell growth
  • Induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities