Learn More
Medchemexpress LLC 3-Bromopyruvic acid | 1113-59-3 | 98.0% | 166.96 | 5 G
SDP
Catalog No. 5000404524
Encompass_Preferred
3-Bromopyruvic acid is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis by interfering with glycolysis. It also induces autophagy by stimulating ROS formation in breast cancer cells, and exhibits antimicrobial activities.
  • Inhibits cell growth and induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities
  • Potent hexokinase (HK)-II inhibitor
  • Shows high tumor selectivity
Catalog No. 50-004-04524 Supplier Medchemexpress LLC Supplier No. HY199925G
3-Bromopyruvic acid is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. It inhibits cell growth and induces apoptosis by interfering with glycolysis. It also induces autophagy by stimulating ROS formation in breast cancer cells, and exhibits antimicrobial activities.
  • Inhibits cell growth and induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities
  • Potent hexokinase (HK)-II inhibitor
  • Shows high tumor selectivity