Learn More
Medchemexpress LLC 3-Bromopyruvic acid | 1113-59-3 | MFCD00002587 | >98.0% | 166.96 | 1 G
SDP
Catalog No. 5000404858
Encompass_Preferred
3-Bromopyruvate (Bromopyruvic acid) is a synthetic analogue of pyruvate and a potent inhibitor of hexokinase II (HK-II), an enzyme frequently overexpressed in cancer cells, contributing to their high glycolytic rates. Its high tumor selectivity allows it to interfere with glycolysis, a primary energy pathway in many cancer cells, thereby inhibiting cell growth and inducing apoptosis. In breast cancer cells, 3-Bromopyruvate has been shown to induce autophagy by stimulating the formation of reactive oxygen species (ROS). Additionally, this compound exhibits significant antimicrobial activities.
  • Potent hexokinase II inhibitor.
  • Exhibits high tumor selectivity.
  • Inhibits cell growth and induces apoptosis.
  • Stimulates reactive oxygen species formation.
  • Demonstrates antimicrobial activities.
Catalog No. 50-004-04858 Supplier Medchemexpress LLC Supplier No. HY199921G
3-Bromopyruvate (Bromopyruvic acid) is a synthetic analogue of pyruvate and a potent inhibitor of hexokinase II (HK-II), an enzyme frequently overexpressed in cancer cells, contributing to their high glycolytic rates. Its high tumor selectivity allows it to interfere with glycolysis, a primary energy pathway in many cancer cells, thereby inhibiting cell growth and inducing apoptosis. In breast cancer cells, 3-Bromopyruvate has been shown to induce autophagy by stimulating the formation of reactive oxygen species (ROS). Additionally, this compound exhibits significant antimicrobial activities.
  • Potent hexokinase II inhibitor.
  • Exhibits high tumor selectivity.
  • Inhibits cell growth and induces apoptosis.
  • Stimulates reactive oxygen species formation.
  • Demonstrates antimicrobial activities.