Learn More
AdipoGen 3-Bromopyruvic acid (5 g)
SDP
Catalog No. 501687444
Encompass_Preferred
3-Bromopyruvic acid (5 g), Adipogen Life Sciences. Chemical. CAS: 1113-59-3. Formula: C3H3BrO3. MW: 167. Synthetic. Cell-permeable pyruvic acid derivative. Hexokinase II (HK2) inhibitor. Useful agent for immunometabolism research. Antitumor agent. Inhibits the rate limiting step of glycolysis and effectively suppress tumor survival in vitro and in vivo. Displays potent cytotoxic activity against cancer cells with mitochondrial respiratory defects and cells in hypoxic environment. Antifungal agent (anticryptococcal). Also used as affinity label for cysteine residues and cross-linker between nucleic acids and proteins.
Catalog No. 50-168-7444 Supplier AdipoGen Supplier No. AGCR13682G005
3-Bromopyruvic acid (5 g), Adipogen Life Sciences. Chemical. CAS: 1113-59-3. Formula: C3H3BrO3. MW: 167. Synthetic. Cell-permeable pyruvic acid derivative. Hexokinase II (HK2) inhibitor. Useful agent for immunometabolism research. Antitumor agent. Inhibits the rate limiting step of glycolysis and effectively suppress tumor survival in vitro and in vivo. Displays potent cytotoxic activity against cancer cells with mitochondrial respiratory defects and cells in hypoxic environment. Antifungal agent (anticryptococcal). Also used as affinity label for cysteine residues and cross-linker between nucleic acids and proteins.