Learn More
Cayman Chemical 3-Bromopyruvic Acid, Purity: ≥95%, CAS: 1113-59-3, 5 g
SDP
Catalog No. 502984368
Encompass_Preferred
An alkylating agent and antimetabolite of pyruvate; decreases proliferation of BEL-7402 cells expressing hexokinase II; reduces tumor growth and induces tumor necrosis in a hepatocellular carcinoma mouse xenograft model at 50 mg/kg per day, six days per week, for three weeks
Catalog No. 50-298-4368 Supplier Cayman Chemical Supplier No. 190685 G
An alkylating agent and antimetabolite of pyruvate; decreases proliferation of BEL-7402 cells expressing hexokinase II; reduces tumor growth and induces tumor necrosis in a hepatocellular carcinoma mouse xenograft model at 50 mg/kg per day, six days per week, for three weeks