Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothieno[3,2-b]thiophene 94.0+%, TCI America™
SDP

Catalog No. B32261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32261G 1 g
B3226200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32261G Supplier TCI America Supplier No. B32261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25121-83-9
Molecular Formula C6H3BrS2
Molecular Weight (g/mol) 219.114
InChI Key YFWCQBIKGWTZQQ-UHFFFAOYSA-N
PubChem CID 14383679
IUPAC Name 6-bromothieno[3,2-b]thiophene
SMILES C1=CSC2=C1SC=C2Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 219.11
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromothieno[3,2-b]thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.