Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3-Bromothiophene, 872-31-1, MFCD00005464, 100g

Catalog No. 500116564
Encompass_Preferred

Molecular Formula C4H3BrS, Purity 97%, Molecular Weight 163.04, Melting Point <25°, Boiling Point 150

Catalog No. 50-011-6564 Supplier Matrix Scientific Supplier No. 004199100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C4H3BrS, Purity 97%, Molecular Weight 163.04, Melting Point <25°, Boiling Point 150

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.