Learn More
Medchemexpress LLC 3-bromothiophene-2-carboxaldehyde | 930-96-1 | 191.05 | 10 G
SDP
Catalog No. 5000448617
Encompass_Preferred
This product is 3-Bromothiophene-2-carboxaldehyde, a biochemical reagent that serves as a biological material or an intermediate. It is intended for research use only by experienced personnel and has not been fully validated for medical applications. The compound should be stored in a well-ventilated area, with the container tightly closed and locked up. Optimal storage conditions are at -20°C, protected from light and stored under nitrogen. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light and under nitrogen. Shipping is at room temperature if the duration is less than 2 weeks.
  • Reagent for biochemical assays
  • Suitable for use as a biological material or intermediate
  • High purity of >99.2%
  • Molecular weight of 191.05
  • Specific storage conditions: -20°C, protected from light, under nitrogen
  • Ships at room temperature for short durations
Catalog No. 50-004-48617 Supplier Medchemexpress LLC Supplier No. HYW00775310G
This product is 3-Bromothiophene-2-carboxaldehyde, a biochemical reagent that serves as a biological material or an intermediate. It is intended for research use only by experienced personnel and has not been fully validated for medical applications. The compound should be stored in a well-ventilated area, with the container tightly closed and locked up. Optimal storage conditions are at -20°C, protected from light and stored under nitrogen. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light and under nitrogen. Shipping is at room temperature if the duration is less than 2 weeks.
  • Reagent for biochemical assays
  • Suitable for use as a biological material or intermediate
  • High purity of >99.2%
  • Molecular weight of 191.05
  • Specific storage conditions: -20°C, protected from light, under nitrogen
  • Ships at room temperature for short durations