Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothiophene-2-sulfonyl chloride, 90%, Thermo Scientific™

Catalog No. MO08006CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08006CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO08006CB Supplier Thermo Scientific Chemicals Supplier No. MO08006CB
Add to Cart

Chemical Identifiers

CAS 170727-02-3
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.532
MDL Number MFCD08059484
InChI Key OPLMNCCLQDVCTP-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo
PubChem CID 13675598
IUPAC Name 3-bromothiophene-2-sulfonyl chloride
SMILES C1=CSC(=C1Br)S(=O)(=O)Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 261.55
Percent Purity 90%
Chemical Name or Material 3-Bromothiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.